Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spoon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258603.2 | NP_001245532.1 | 4 - 5 | 1956 | 92 | |
NM_131993.4 | NP_572221.1 | 4 - 5 | 2118 | 92 | ||
NM_167033.3 | NP_726991.1 | 4 - 5 | 2445 | 92 | ||
NM_167034.2 | NP_726992.2 | 4 - 5 | 1845 | 92 | ||
GenBank mRNA | AY051641.1 | - | 4 - 5 | 2091 | 92 | |
BT099605.1 | - | 4 - 5 | 1841 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |