Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132007.4 | NP_572235.1 | 1 - 2 | 246 | 76 | |
NM_167040.4 | NP_727007.1 | 1 - 2 | 246 | 76 | ||
GenBank mRNA | AY050239.1 | - | 1 - 2 | 196 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |