Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132057.3 | NP_572285.2 | 3 - 4 | 795 | 67 | |
GenBank mRNA | BT044143.1 | - | 3 - 4 | 794 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |