Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042904.4 | NP_001036369.3 | 19 - 20 | 7657 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |