Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042904.4 | NP_001036369.3 | 8 - 9 | 3897 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |