Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elfless | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169532.1 | NP_001163003.1 | 3 - 4 | 425 | 72 | |
NM_136016.4 | NP_609860.2 | 3 - 4 | 425 | 72 | ||
GenBank mRNA | BT072897.1 | - | 3 - 4 | 425 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |