Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103707.2 | NP_001097177.1 | 14 - 15 | 3856 | 66 | |
NM_001201908.2 | NP_001188837.1 | 14 - 15 | 3550 | 66 | ||
NM_001201909.2 | NP_001188838.1 | 14 - 15 | 3856 | 66 | ||
NM_001299115.1 | NP_001286044.1 | 14 - 15 | 3723 | 66 | ||
NM_001299116.1 | NP_001286045.1 | 15 - 16 | 2943 | 66 | ||
NM_136041.2 | NP_609885.2 | 14 - 15 | 3609 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |