Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144363.2 | NP_001137835.2 | 3 - 4 | 1045 | 64 | |
NM_136059.4 | NP_609903.2 | 3 - 4 | 1033 | 64 | ||
GenBank mRNA | AJ344347.1 | - | 3 - 4 | 820 | 64 | |
BT024418.1 | - | 3 - 4 | 1013 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |