Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299140.1 | NP_001286069.1 | 2 - 3 | 2945 | 102 | |
NM_136073.3 | NP_609917.2 | 2 - 3 | 2951 | 102 | ||
GenBank mRNA | BT021951.1 | - | 2 - 3 | 2935 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |