Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299140.1 | NP_001286069.1 | 7 - 8 | 5067 | 91 | |
NM_136073.3 | NP_609917.2 | 7 - 8 | 5073 | 91 | ||
GenBank mRNA | BT021951.1 | - | 7 - 8 | 5057 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |