Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138984.4 | NP_620469.2 | 5 - 6 | 2039 | 57 | |
GenBank mRNA | BT053678.1 | - | 5 - 6 | 2037 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |