Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169916.2 | NP_001163387.2 | 2 - 3 | 409 | 72 | |
NM_001274670.1 | NP_001261599.1 | 2 - 3 | 933 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |