Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meso18E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169328.2 | NP_001162799.1 | 3 - 4 | 1980 | 77 | |
NM_001298531.1 | NP_001285460.1 | 4 - 5 | 1472 | 77 | ||
NM_001298532.1 | NP_001285461.1 | 3 - 4 | 1503 | 77 | ||
NM_144358.3 | NP_652615.1 | 3 - 4 | 1405 | 77 | ||
GenBank mRNA | AJ271814.1 | - | 3 - 4 | 1933 | 77 | |
AY058570.1 | - | 3 - 4 | 1358 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |