Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01797431_g1
See other SHOT GE Assays ›
Gene Symbol:
shot
Gene Name:
short stop
Gene Aliases:
42/4, CG18076, CG18637, Dmel\CG18076, Dmel_CG18076, Grv, Kak, Shot, Su(Mir)1, grv, kak, kakp, kop, l(2)CA4, l(2)k03010, l(2)k04204, l(2)k05434, l(2)k05821, l(2)k10821, l(2)k15606, shortstop/kakapo
Chromosome Location:
Chr.2R: 13864237 - 13942110 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0013733
Assay ID
Dm01797431_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: shot
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001144185.2
NP_001137657.1
8 - 9
2479
71
NM_001144186.2
NP_001137658.1
8 - 9
2479
71
NM_001144187.2
NP_001137659.1
8 - 9
2479
71
NM_001144188.2
NP_001137660.1
8 - 9
2702
71
NM_001144189.2
NP_001137661.1
8 - 9
2702
71
NM_001169676.1
NP_001163147.1
5 - 6
2495
71
NM_001169677.1
NP_001163148.1
8 - 9
2479
71
NM_001169678.1
NP_001163149.1
8 - 9
2702
71
NM_001169679.2
NP_001163150.1
5 - 6
2495
71
NM_001259378.1
NP_001246307.1
5 - 6
2495
71
NM_001259379.1
NP_001246308.1
5 - 6
2495
71
NM_001259380.1
NP_001246309.1
9 - 10
2497
71
NM_001259381.1
NP_001246310.1
9 - 10
2539
71
NM_001259383.1
NP_001246312.1
5 - 6
2495
71
NM_001259384.1
NP_001246313.1
5 - 6
2495
71
NM_079009.3
NP_523733.2
8 - 9
2702
71
NM_166015.2
NP_725335.1
8 - 9
2479
71
NM_166016.2
NP_725336.1
8 - 9
2479
71
NM_166017.2
NP_725337.1
8 - 9
2702
71
NM_166018.3
NP_725338.1
5 - 6
2495
71
NM_166019.3
NP_725339.1
5 - 6
2495
71
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Drosophila melanogaster
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
microtubule-based process
negative regulation of microtubule depolymerization
dorsal closure
axonogenesis
open tracheal system development
axon midline choice point recognition
mushroom body development
actin cytoskeleton organization
regulation of axon extension
oocyte fate determination
cytoplasmic microtubule organization
axoneme assembly
branch fusion, open tracheal system
lumen formation, open tracheal system
wound healing
cortical microtubule organization
intermediate filament cytoskeleton organization
positive regulation of axon extension
dendrite morphogenesis
positive regulation of filopodium assembly
actin binding
structural molecule activity
calcium ion binding
protein binding
microtubule binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0