Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299482.1 | NP_001286411.1 | 7 - 8 | 1752 | 84 | |
NM_001299483.1 | NP_001286412.1 | 6 - 7 | 1635 | 84 | ||
NM_001299484.1 | NP_001286413.1 | 6 - 7 | 1635 | 84 | ||
NM_137097.3 | NP_610941.1 | 6 - 7 | 1635 | 84 | ||
NM_166040.2 | NP_725369.1 | 7 - 8 | 1752 | 84 | ||
GenBank mRNA | AY058366.1 | - | 6 - 7 | 1609 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |