Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299498.1 | NP_001286427.1 | 1 - 2 | 815 | 70 | |
NM_166063.2 | NP_725407.1 | 1 - 2 | 815 | 70 | ||
GenBank mRNA | AY084178.1 | - | 1 - 2 | 814 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |