Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: row | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274063.2 | NP_001260992.1 | 2 - 3 | 1860 | 84 | |
NM_137175.4 | NP_611019.2 | 2 - 3 | 1860 | 84 | ||
NM_166081.3 | NP_725441.1 | 2 - 3 | 1860 | 84 | ||
GenBank mRNA | AY061301.1 | - | 2 - 3 | 1829 | 84 | |
BT016036.1 | - | 2 - 3 | 1839 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |