Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166144.2 | NP_725530.1 | 4 - 5 | 1752 | 67 | |
NM_166146.2 | NP_725532.1 | 4 - 5 | 1752 | 67 | ||
GenBank mRNA | AF212366.1 | - | 4 - 5 | 1715 | 67 | |
AF212368.1 | - | 4 - 5 | 1715 | 67 | ||
BT032842.1 | - | 4 - 5 | 1714 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |