Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259933.1 | NP_001246862.1 | 2 - 3 | 1333 | 59 | |
NM_001259934.1 | NP_001246863.1 | 2 - 3 | 1189 | 59 | ||
NM_079471.3 | NP_524195.2 | 1 - 2 | 1474 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |