Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AcCoAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014599.2 | NP_001014599.2 | 5 - 6 | 1758 | 67 | |
NM_079472.3 | NP_524196.2 | 5 - 6 | 1402 | 67 | ||
NM_168894.3 | NP_730611.1 | 5 - 6 | 2020 | 67 | ||
GenBank mRNA | BT126330.1 | - | 5 - 6 | 2020 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |