Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202222.1 | NP_001189151.1 | 2 - 3 | 128 | 69 | |
NM_001202223.1 | NP_001189152.1 | 1 - 2 | 121 | 69 | ||
NM_079479.5 | NP_524203.2 | 2 - 3 | 291 | 69 | ||
NM_168907.3 | NP_730636.1 | 2 - 3 | 261 | 69 | ||
NM_168908.3 | NP_730637.1 | 2 - 3 | 306 | 69 | ||
NM_206415.3 | NP_996137.1 | 2 - 3 | 291 | 69 | ||
GenBank mRNA | AY058317.1 | - | 2 - 3 | 260 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |