Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275277.1 | NP_001262206.1 | 5 - 6 | 707 | 84 | |
NM_001275278.1 | NP_001262207.1 | 5 - 6 | 707 | 84 | ||
NM_001300215.1 | NP_001287144.1 | 5 - 6 | 893 | 84 | ||
NM_168949.2 | NP_730713.1 | 5 - 6 | 707 | 84 | ||
GenBank mRNA | BT100236.1 | - | 5 - 6 | 685 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |