Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IleRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080101.4 | NP_524840.2 | 5 - 6 | 2792 | 139 | |
NM_168952.4 | NP_730716.1 | 4 - 5 | 2871 | 139 | ||
NM_176387.2 | NP_788565.1 | 5 - 6 | 2817 | 139 | ||
GenBank mRNA | AY118538.1 | - | 4 - 5 | 2861 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |