Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038924.4 | NP_001034013.2 | 3 - 4 | 1230 | 75 | |
NM_001038925.3 | NP_001034014.1 | 4 - 5 | 1279 | 75 | ||
NM_001038926.2 | NP_001034015.1 | 4 - 5 | 1345 | 75 | ||
NM_001274917.1 | NP_001261846.1 | 4 - 5 | 1345 | 75 | ||
NM_206355.4 | NP_996077.1 | 4 - 5 | 1345 | 75 | ||
NM_206356.3 | NP_996078.1 | 4 - 5 | 1345 | 75 | ||
NM_206357.3 | NP_996079.1 | 4 - 5 | 1703 | 75 | ||
NM_206358.4 | NP_996080.1 | 4 - 5 | 1520 | 75 | ||
NM_206359.4 | NP_996081.1 | 4 - 5 | 1524 | 75 | ||
NM_206360.1 | NP_996082.1 | 5 - 6 | 1648 | 75 | ||
GenBank mRNA | AY069651.1 | - | 4 - 5 | 1503 | 75 |
Gene Symbol: CG42507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169972.2 | NP_001163443.1 | 4 - 5 | 1345 | 75 | |
NM_001169973.2 | NP_001163444.1 | 4 - 5 | 1345 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster Inventoried |