Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CrebA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079363.5 | NP_524087.3 | 3 - 4 | 1772 | 59 | |
NM_206374.2 | NP_996096.1 | 3 - 4 | 1772 | 59 | ||
GenBank mRNA | BT025219.1 | - | 3 - 4 | 1770 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |