Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259860.2 | NP_001246789.1 | 3 - 4 | 3606 | 89 | |
NM_206380.2 | NP_996102.1 | 3 - 4 | 3452 | 89 | ||
NM_206381.1 | NP_996103.1 | 2 - 3 | 3702 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |