Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259888.3 | NP_001246817.1 | 2 - 3 | 446 | 75 | |
NM_080119.6 | NP_524858.1 | 3 - 4 | 612 | 75 | ||
NM_206396.4 | NP_996118.1 | 2 - 3 | 511 | 75 | ||
GenBank mRNA | AY118552.1 | - | 2 - 3 | 474 | 75 | |
X56689.1 | - | 597 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |