Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259940.1 | NP_001246869.1 | 4 - 5 | 721 | 69 | |
NM_206416.2 | NP_996138.2 | 4 - 5 | 721 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |