Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130731.3 | NP_570087.1 | 2 - 3 | 632 | 77 | |
NM_206623.2 | NP_996346.2 | 2 - 3 | 632 | 77 | ||
NM_206624.2 | NP_996347.1 | 2 - 3 | 342 | 77 | ||
NM_206625.2 | NP_996348.1 | 2 - 3 | 632 | 77 | ||
GenBank mRNA | AY060395.1 | - | 2 - 3 | 603 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |