Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132092.3 | NP_572320.2 | 8 - 9 | 2753 | 67 | |
NM_206634.2 | NP_996357.1 | 7 - 8 | 2741 | 67 | ||
NM_206635.3 | NP_996358.2 | 7 - 8 | 2580 | 67 | ||
GenBank mRNA | BT010027.1 | - | 7 - 8 | 2741 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |