Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038746.3 | NP_001033835.2 | 16 - 17 | 7288 | 72 | |
NM_001038747.3 | NP_001033836.1 | 11 - 12 | 3308 | 72 | ||
NM_001042798.2 | NP_001036263.1 | 9 - 10 | 2843 | 72 | ||
NM_001258645.1 | NP_001245574.1 | 17 - 18 | 5232 | 72 | ||
NM_001258646.1 | NP_001245575.1 | 7 - 8 | 2616 | 72 | ||
NM_001258647.1 | NP_001245576.1 | 18 - 19 | 6571 | 72 | ||
NM_001272397.1 | NP_001259326.1 | 16 - 17 | 6931 | 72 | ||
NM_001272398.1 | NP_001259327.1 | 11 - 12 | 4476 | 72 | ||
NM_001298081.1 | NP_001285010.1 | 15 - 16 | 6046 | 72 | ||
NM_001298082.1 | NP_001285011.1 | 14 - 15 | 4747 | 72 | ||
NM_132235.5 | NP_572463.1 | 12 - 13 | 4607 | 72 | ||
NM_132236.6 | NP_572464.4 | 11 - 12 | 4040 | 72 | ||
NM_206652.2 | NP_996375.1 | 7 - 8 | 2664 | 72 | ||
NM_206653.3 | NP_996376.1 | 11 - 12 | 4269 | 72 | ||
GenBank mRNA | BT010281.1 | - | 11 - 12 | 3306 | 72 | |
BT099747.1 | - | 7 - 8 | 2664 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |