Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038746.3 | NP_001033835.2 | 3 - 4 | 1694 | 60 | |
NM_001258645.1 | NP_001245574.1 | 4 - 5 | 1117 | 60 | ||
NM_001258647.1 | NP_001245576.1 | 4 - 5 | 1214 | 60 | ||
NM_001272397.1 | NP_001259326.1 | 3 - 4 | 1694 | 60 | ||
NM_001298081.1 | NP_001285010.1 | 3 - 4 | 1694 | 60 | ||
NM_001298082.1 | NP_001285011.1 | 3 - 4 | 1694 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |