Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Graf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298367.1 | NP_001285296.1 | 11 - 12 | 3359 | 78 | |
NM_132842.3 | NP_573070.2 | 11 - 12 | 3332 | 78 | ||
NM_206718.2 | NP_996441.2 | 12 - 13 | 3644 | 78 | ||
NM_206719.2 | NP_996442.1 | 11 - 12 | 3559 | 78 | ||
NM_206720.2 | NP_996443.1 | 12 - 13 | 3617 | 78 | ||
NM_206721.2 | NP_996444.2 | 11 - 12 | 3586 | 78 | ||
NM_206722.2 | NP_996445.1 | 11 - 12 | 3242 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |