Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01799454_g1
See other SD GE Assays ›
Gene Symbol:
sd
Gene Name:
scalloped
Gene Aliases:
CG8544, DROEXO, DROEXO2, Dmel\CG8544, Dmel_CG8544, EP(X)1088, EP1088, SD, Sd, TEAD, anon-EST:Liang-2.14, clone 2.14, l(1)G0239, l(1)G0262, l(1)G0309, l(1)G0315, l(1)G0483, l(1)HF394, l(1)III, sp
Chromosome Location:
Chr.X: 15804370 - 15827682 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003345
Assay ID
Dm01799454_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: sd
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001103519.3
NP_001096989.1
9 - 10
1900
99
NM_001103520.3
NP_001096990.1
10 - 11
1864
99
NM_001103521.3
NP_001096991.1
10 - 11
1659
99
NM_001103522.3
NP_001096992.1
10 - 11
1751
99
NM_001169290.2
NP_001162761.1
10 - 11
1849
99
NM_001169291.2
NP_001162762.1
10 - 11
1892
99
NM_001258767.2
NP_001245696.1
10 - 11
2657
99
NM_001258768.1
NP_001245697.1
10 - 11
1725
99
NM_001258769.2
NP_001245698.1
9 - 10
1139
99
NM_001258770.3
NP_001245699.1
10 - 11
1984
99
NM_001258771.3
NP_001245700.1
9 - 10
1197
99
NM_001258772.2
NP_001245701.1
10 - 11
2393
99
NM_001258773.2
NP_001245702.1
10 - 11
1967
99
NM_001272652.1
NP_001259581.1
10 - 11
2282
99
NM_001272653.2
NP_001259582.1
11 - 12
1915
99
NM_001272654.2
NP_001259583.1
10 - 11
2003
99
NM_001298370.1
NP_001285299.1
9 - 10
1278
99
NM_001316637.1
NP_001303566.1
10 - 11
2657
99
NM_001382196.1
NP_001368984.1
10 - 11
1984
99
NM_078614.5
NP_511169.1
11 - 12
1806
99
NM_167465.3
NP_727900.1
9 - 10
1289
99
NM_206727.3
NP_996450.1
9 - 10
1453
99
GenBank mRNA
BT133072.1
-
10 - 11
1846
99
M83787.1
-
11 - 12
1804
99
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Drosophila melanogaster
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
compound eye morphogenesis
negative regulation of neuroblast proliferation
sensory organ development
imaginal disc-derived wing morphogenesis
imaginal disc-derived leg morphogenesis
somatic muscle development
wing disc development
hippo signaling
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic organ development
cardiac muscle cell development
stem cell proliferation
regulation of heart morphogenesis
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription corepressor binding
transcription coactivator binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
protein domain specific binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0