Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103523.2 | NP_001096993.1 | 3 - 4 | 616 | 85 | |
NM_001103524.2 | NP_001096994.1 | 3 - 4 | 510 | 85 | ||
NM_001298371.1 | NP_001285300.1 | 4 - 5 | 721 | 85 | ||
NM_057694.3 | NP_477042.1 | 3 - 4 | 629 | 85 | ||
NM_167466.2 | NP_727901.1 | 3 - 4 | 586 | 85 | ||
NM_206728.2 | NP_996451.1 | 3 - 4 | 726 | 85 | ||
NM_206729.2 | NP_996452.1 | 3 - 4 | 667 | 85 | ||
GenBank mRNA | AY119615.1 | - | 3 - 4 | 613 | 85 | |
Z14133.1 | - | 543 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |