Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169292.2 | NP_001162763.1 | 3 - 4 | 1295 | 72 | |
NM_132855.4 | NP_573083.2 | 3 - 4 | 1385 | 72 | ||
NM_206730.3 | NP_996453.1 | 3 - 4 | 1396 | 72 | ||
GenBank mRNA | BT021420.1 | - | 3 - 4 | 1356 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |