Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042813.2 | NP_001036278.1 | 13 - 14 | 2887 | 79 | |
NM_001042814.2 | NP_001036279.1 | 13 - 14 | 2899 | 79 | ||
NM_001169295.1 | NP_001162766.1 | 13 - 14 | 2758 | 79 | ||
NM_001169296.1 | NP_001162767.1 | 12 - 13 | 2474 | 79 | ||
NM_001169297.2 | NP_001162768.1 | 13 - 14 | 2776 | 79 | ||
NM_001272659.2 | NP_001259588.1 | 14 - 15 | 2626 | 79 | ||
NM_001272660.2 | NP_001259589.1 | 14 - 15 | 2747 | 79 | ||
NM_001298377.1 | NP_001285306.1 | 14 - 15 | 2581 | 79 | ||
NM_080114.4 | NP_524853.2 | 12 - 13 | 2456 | 79 | ||
NM_167470.3 | NP_727910.1 | 13 - 14 | 2678 | 79 | ||
NM_167471.5 | NP_727911.1 | 13 - 14 | 2758 | 79 | ||
NM_206742.1 | NP_996465.1 | 13 - 14 | 2568 | 79 | ||
NM_206743.2 | NP_996466.1 | 13 - 14 | 2569 | 79 | ||
NM_206744.3 | NP_996467.1 | 13 - 14 | 2569 | 79 | ||
NM_206745.3 | NP_996468.1 | 13 - 14 | 2568 | 79 | ||
GenBank mRNA | BT010049.1 | - | 13 - 14 | 2735 | 79 | |
X59435.1 | - | 2474 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |