Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042813.2 | NP_001036278.1 | 8 - 9 | 2139 | 71 | |
NM_001042814.2 | NP_001036279.1 | 8 - 9 | 2151 | 71 | ||
NM_001169295.1 | NP_001162766.1 | 8 - 9 | 2010 | 71 | ||
NM_001169296.1 | NP_001162767.1 | 7 - 8 | 1708 | 71 | ||
NM_001169297.2 | NP_001162768.1 | 8 - 9 | 2010 | 71 | ||
NM_001272660.2 | NP_001259589.1 | 8 - 9 | 1987 | 71 | ||
NM_001298377.1 | NP_001285306.1 | 8 - 9 | 1821 | 71 | ||
NM_080114.4 | NP_524853.2 | 7 - 8 | 1708 | 71 | ||
NM_167470.3 | NP_727910.1 | 8 - 9 | 1930 | 71 | ||
NM_167471.5 | NP_727911.1 | 8 - 9 | 2010 | 71 | ||
NM_206742.1 | NP_996465.1 | 8 - 9 | 1820 | 71 | ||
NM_206743.2 | NP_996466.1 | 8 - 9 | 1821 | 71 | ||
NM_206744.3 | NP_996467.1 | 8 - 9 | 1821 | 71 | ||
NM_206745.3 | NP_996468.1 | 8 - 9 | 1820 | 71 | ||
GenBank mRNA | BT010049.1 | - | 8 - 9 | 1987 | 71 | |
X59435.1 | - | 1708 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |