Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258774.2 | NP_001245703.1 | 5 - 6 | 1116 | 107 | |
NM_078633.4 | NP_523357.2 | 5 - 6 | 1052 | 107 | ||
NM_206747.3 | NP_996470.2 | 5 - 6 | 1052 | 107 | ||
GenBank mRNA | AF328906.1 | - | 4 - 5 | 1008 | 107 | |
AY069686.1 | - | 5 - 6 | 1036 | 107 | ||
BT133089.1 | - | 5 - 6 | 1030 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |