Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144728.3 | NP_001138200.1 | 12 - 13 | 2292 | 61 | |
NM_001258775.3 | NP_001245704.1 | 12 - 13 | 2292 | 61 | ||
NM_001258776.2 | NP_001245705.1 | 13 - 14 | 2283 | 61 | ||
NM_001272661.2 | NP_001259590.1 | 13 - 14 | 2491 | 61 | ||
NM_001272662.2 | NP_001259591.1 | 13 - 14 | 2491 | 61 | ||
NM_001298382.1 | NP_001285311.1 | 12 - 13 | 2974 | 61 | ||
NM_078634.7 | NP_523358.5 | 12 - 13 | 2974 | 61 | ||
NM_206752.3 | NP_996475.2 | 12 - 13 | 2292 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |