Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169302.2 | NP_001162773.1 | 5 - 6 | 1526 | 82 | |
NM_001169303.2 | NP_001162774.1 | 6 - 7 | 1454 | 82 | ||
NM_078644.4 | NP_523368.2 | 5 - 6 | 1526 | 82 | ||
NM_167506.3 | NP_727963.1 | 5 - 6 | 1535 | 82 | ||
NM_167507.3 | NP_727964.1 | 5 - 6 | 1635 | 82 | ||
NM_167508.2 | NP_727965.1 | 5 - 6 | 1569 | 82 | ||
NM_167509.3 | NP_727966.1 | 5 - 6 | 1629 | 82 | ||
NM_167510.3 | NP_727967.1 | 6 - 7 | 1746 | 82 | ||
NM_206763.2 | NP_996486.1 | 5 - 6 | 1513 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |