Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169317.1 | NP_001162788.1 | 5 - 6 | 826 | 143 | |
NM_001201739.2 | NP_001188668.1 | 7 - 8 | 3118 | 143 | ||
NM_001258797.1 | NP_001245726.1 | 6 - 7 | 1761 | 143 | ||
NM_001258798.2 | NP_001245727.1 | 9 - 10 | 2725 | 143 | ||
NM_001272735.3 | NP_001259664.1 | 7 - 8 | 3118 | 143 | ||
NM_001272736.2 | NP_001259665.2 | 6 - 7 | 1306 | 143 | ||
NM_001272737.1 | NP_001259666.2 | 6 - 7 | 2081 | 143 | ||
NM_001316640.1 | NP_001303569.1 | 6 - 7 | 1371 | 143 | ||
NM_078669.5 | NP_523393.3 | 6 - 7 | 1371 | 143 | ||
NM_167592.4 | NP_728120.1 | 6 - 7 | 905 | 143 | ||
NM_167594.4 | NP_728122.1 | 7 - 8 | 3118 | 143 | ||
NM_167595.4 | NP_728123.1 | 6 - 7 | 1371 | 143 | ||
NM_167596.5 | NP_728124.1 | 6 - 7 | 905 | 143 | ||
NM_206774.2 | NP_996497.1 | 6 - 7 | 1314 | 143 | ||
NM_206775.5 | NP_996498.2 | 9 - 10 | 2725 | 143 | ||
GenBank mRNA | BT050432.1 | - | 6 - 7 | 905 | 143 | |
X06184.1 | - | 824 | 143 | |||
X07132.1 | - | 999 | 143 | |||
X07133.1 | - | 703 | 143 | |||
X07134.1 | - | 1314 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |