Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169342.2 | NP_001162813.1 | 2 - 3 | 1199 | 101 | |
NM_057389.6 | NP_476737.3 | 2 - 3 | 1199 | 101 | ||
NM_057390.6 | NP_476738.3 | 2 - 3 | 1199 | 101 | ||
NM_206801.3 | NP_996524.2 | 2 - 3 | 760 | 101 | ||
GenBank mRNA | BT133439.1 | - | 2 - 3 | 1149 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |