Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298579.1 | NP_001285508.1 | 3 - 4 | 1360 | 85 | |
NM_134580.3 | NP_608424.1 | 3 - 4 | 1483 | 85 | ||
NM_167726.2 | NP_728384.1 | 3 - 4 | 1562 | 85 | ||
GenBank mRNA | AY058508.1 | - | 3 - 4 | 1334 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |