Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134613.3 | NP_608457.1 | 1 | 1277 | 63 | |
GenBank mRNA | AY071245.1 | - | 1 | 1228 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |