Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tilB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134616.3 | NP_608460.1 | 3 - 4 | 996 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |