Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rempA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134686.2 | NP_608530.2 | 5 - 6 | 4772 | 75 | |
NM_205886.2 | NP_995608.1 | 5 - 6 | 4488 | 75 | ||
GenBank mRNA | BT021360.1 | - | 5 - 6 | 4772 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |