Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134713.4 | NP_608557.4 | 3 - 4 | 416 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |