Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272923.2 | NP_001259852.1 | 1 - 2 | 514 | 63 | |
NM_134719.4 | NP_608563.1 | 1 - 2 | 514 | 63 | ||
GenBank mRNA | AY058573.1 | - | 1 - 2 | 496 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |