Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134730.3 | NP_608574.2 | 3 - 4 | 439 | 54 | |
GenBank mRNA | BT025234.2 | - | 3 - 4 | 435 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |